Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CFTR inhibitor-172 98%

Supplier: AMBEED, INC
Danger

A285038-10MG A285038-1MG A285038-25MG A285038-5MG
77277-046EA 34 USD InStock
77277-046 77277-048 77277-050 77277-052
CFTR inhibitor-172 98%
CFTR inhibitor-172
Precaución: for Research Use Only


Formula: C₁₈H₁₀F₃NO₃S₂
MDL Number: MFCD01164609
CAS Number: 307510-92-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty