Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A285458-10G A285458-1G A285458-25G A285458-5G
77277-060EA 34 USD InStock
77277-060 77277-062 77277-064 77277-066
2-Phenoxyphenylboronic acid 98%
2-Phenoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₁BO₃
MDL Number: MFCD01001592
CAS Number: 108238-09-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty