Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A286076-100MG A286076-1G A286076-250MG A286076-5G
77289-740EA 11 USD InStock
77289-740 77289-742 77289-744 77289-746
Methyl-4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride 98%
Methyl-4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₁ClN₂O₂
MDL Number: MFCD01318761
CAS Number: 180258-45-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty