Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-o-toluonitrile 97%

Supplier: AMBEED, INC
Warning

A287653-100G A287653-10G A287653-1G A287653-25G A287653-5G
77277-116EA 56 USD InStock
77277-116 77277-118 77277-120 77277-122 77277-124
5-Nitro-o-toluonitrile 97%
5-Nitro-o-toluonitrile
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
MW: 162.15
Boiling Pt: 174
MDL Number: MFCD00017040
CAS Number: 939-83-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty