Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(3-bromopropyl)benzene 95%

Supplier: AMBEED, INC
Warning

A288213-250MG A288213-100MG A288213-10G A288213-1G
77239-008EA 31 USD InStock
77239-008 77746-531 77746-532 77746-533
1-Bromo-4-(3-bromopropyl)benzene 95%
1-Bromo-4-(3-bromopropyl)benzene
Precaución: for Research Use Only


Formula: C₉H₁₀Br₂
MW: 277.984 g/mol
MDL Number: MFCD09744416
CAS Number: 90562-10-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty