Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxy-4-formylphenyl acetate 97%

Supplier: AMBEED, INC
Warning

A288840-100G A288840-10G A288840-250MG A288840-25G A288840-5G
77262-784EA 50 USD InStock
77262-784 77262-786 77262-788 77262-790 77262-792
2-Ethoxy-4-formylphenyl acetate 97%
2-Ethoxy-4-formylphenyl acetate
Precaución: for Research Use Only


Formula: C₁₁H₁₂O₄
MDL Number: MFCD01666413
CAS Number: 72207-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty