Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxybenzenecarboximidamide 95%

Supplier: AMBEED, INC
Danger

A290671-100MG A290671-1MG A290671-250MG A290671-1G
77289-784EA 46 USD InStock
77289-784 77289-786 77289-788 77748-564
4-Methoxybenzenecarboximidamide 95%
4-Methoxybenzenecarboximidamide
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O
MDL Number: MFCD00029547
CAS Number: 22265-37-8
UN: 3263
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty