Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Leukadherin-1 98%

Supplier: AMBEED, INC
Danger

A292458-100MG A292458-10MG A292458-250MG A292458-50MG A292458-5MG
77262-838EA 257 USD InStock
77262-838 77262-840 77262-842 77262-844 77262-846 77262-848 77262-850
Leukadherin-1 98%
Leukadherin-1
Precaución: for Research Use Only


Formula: C₂₂H₁₅NO₄S₂
MW: 421.485 g/mol
MDL Number: MFCD01151276
CAS Number: 344897-95-6
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty