Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-methoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A292921-1G A292921-25G A292921-5G
77277-182EA 11 USD InStock
77277-182 77277-184 77277-186
3-Bromo-5-methoxyphenylboronic acid 98%
3-Bromo-5-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BBrO₃
MDL Number: MFCD07369759
CAS Number: 849062-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty