Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodosalicylic acid 95%

Supplier: AMBEED, INC
Danger

A292976-10G A292976-1G A292976-5G
77262-860EA 50 USD InStock
77262-860 77262-862 77262-864
4-Iodosalicylic acid 95%
4-Iodosalicylic acid
Precaución: for Research Use Only


Formula: C₇H₅IO₃
MDL Number: MFCD00061127
CAS Number: 16870-28-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty