Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hexyn-1-ol 95%

Supplier: AMBEED, INC
Warning

A293215-1G A293215-250MG A293215-5G
77277-202EA 12 USD InStock
77277-202 77277-204 77277-206
3-Hexyn-1-ol 95%
3-Hexyn-1-ol
Precaución: for Research Use Only


Formula: C₆H₁₀O
MDL Number: MFCD00002957
CAS Number: 1002-28-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty