Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Prolinamide 97%

Supplier: AMBEED, INC
Warning

A293623-100G A293623-10G A293623-25G A293623-500G A293623-5G A293623-1000G
77289-834EA 63 USD InStock
77289-834 77289-836 77289-838 77289-840 77289-842 77748-573
L-Prolinamide 97%
L-Prolinamide
Precaución: for Research Use Only


Formula: C₅H₁₀N₂O
MDL Number: MFCD00005253
CAS Number: 7531-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty