Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-acetamido-2-ethoxybenzoate 99%

Supplier: AMBEED, INC
Warning

A294987-100G A294987-25G
77277-244EA 182 USD InStock
77277-244 77277-246
Methyl 4-acetamido-2-ethoxybenzoate 99%
Methyl 4-acetamido-2-ethoxybenzoate
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00075809
CAS Number: 59-06-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty