Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3,5-Bis(trifluoromethyl)-1H-pyrazol-1-yl)phenylamine 98%

Supplier: AMBEED, INC
Warning

A295487-100MG A295487-1G A295487-250MG A295487-5G
77289-870EA 76 USD InStock
77289-870 77289-872 77289-874 77289-876
4-(3,5-Bis(trifluoromethyl)-1H-pyrazol-1-yl)phenylamine 98%
4-(3,5-Bis(trifluoromethyl)-1H-pyrazol-1-yl)phenylamine
Precaución: for Research Use Only


Formula: C₁₁H₇F₆N₃
MW: 295.1838 g/mol
MDL Number: MFCD00220969
CAS Number: 123066-64-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty