Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Aminoethyl)-5-(3-(4-butoxyphenyl)propylidene)thiazolidine-2,4-dione hydrochloride 98%

Supplier: AMBEED, INC
Warning

A295989-100MG A295989-10MG A295989-1MG A295989-25MG A295989-50MG A295989-5MG
77239-152EA 289 USD InStock
77239-152 77239-154 77239-156 77239-158 77239-160 77239-162
3-(2-Aminoethyl)-5-(3-(4-butoxyphenyl)propylidene)thiazolidine-2,4-dione hydrochloride 98%
3-(2-Aminoethyl)-5-(3-(4-butoxyphenyl)propylidene)thiazolidine-2,4-dione hydrochloride
Precaución: for Research Use Only


Formula: C₁₈H₂₅ClN₂O₃S
MDL Number: MFCD28160629
CAS Number: 1449240-68-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty