Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1'H-2,2'-Bibenzo[d]imidazole 97%

Supplier: AMBEED, INC
Warning

A297570-100MG A297570-1G A297570-250MG A297570-5G
77239-190EA 6 USD InStock
77239-190 77239-192 77239-194 77239-196
1H,1'H-2,2'-Bibenzo[d]imidazole 97%
1H,1'H-2,2'-Bibenzo[d]imidazole
Precaución: for Research Use Only


Formula: C₁₄H₁₀N₄
MDL Number: MFCD00047158
CAS Number: 6965-02-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty