Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-fluoroquinoline 97%

Supplier: AMBEED, INC
Warning

A298367-100G A298367-10G A298367-1G A298367-250MG A298367-25G
77262-934EA 177 USD InStock
77262-934 77262-936 77262-938 77262-940 77262-942
4-Chloro-6-fluoroquinoline 97%
4-Chloro-6-fluoroquinoline
Precaución: for Research Use Only


Formula: C₉H₅ClFN
MW: 181.60 g/mol
MDL Number: MFCD00278783
CAS Number: 391-77-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty