Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-1-pentanol 99%

Supplier: AMBEED, INC
Danger

A299156-10G A299156-1G A299156-25G A299156-5G
77289-924EA 57 USD InStock
77289-924 77289-926 77289-928 77289-930 77289-932
4-Methyl-1-pentanol 99%
4-Methyl-1-pentanol
Precaución: for Research Use Only


Formula: C₆H₁₄O
MDL Number: MFCD00002962
CAS Number: 626-89-1
UN: 1987
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty