Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Chloro-6'-fluoroacetophenone 97%

Supplier: AMBEED, INC
Warning

A299625-100G A299625-10G A299625-1G A299625-25G A299625-5G
77239-228EA 203 USD InStock
77239-228 77239-230 77239-232 77239-234 77239-236
2'-Chloro-6'-fluoroacetophenone 97%
2'-Chloro-6'-fluoroacetophenone
Precaución: for Research Use Only


Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD01631390
CAS Number: 87327-69-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty