Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluorophenol 98%

Supplier: AMBEED, INC
Warning

A300724-100G A300724-10G A300724-25G A300724-500G A300724-5G
77289-974EA 86 USD InStock
77289-974 77289-976 77289-978 77289-980 77289-982
4-Chloro-2-fluorophenol 98%
4-Chloro-2-fluorophenol
Precaución: for Research Use Only


Formula: C₆H₄ClFO
MDL Number: MFCD00075030
CAS Number: 348-62-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty