Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-1-fluoro-4-nitrobenzene 98%

Supplier: AMBEED, INC
Warning

A300938-100G A300938-10G A300938-1G A300938-25G A300938-5G
77289-984EA 99 USD InStock
77289-984 77289-986 77289-988 77289-990 77289-992
3-Chloro-1-fluoro-4-nitrobenzene 98%
3-Chloro-1-fluoro-4-nitrobenzene
Precaución: for Research Use Only


Formula: C₆H₃ClFNO₂
MDL Number: MFCD03412200
CAS Number: 2106-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty