Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alvelestat 98+%

Supplier: AMBEED, INC
Warning

A301653-10MG A301653-1MG A301653-2MG A301653-5MG A301653-50MG
77239-254EA 42 USD InStock
77239-254 77239-256 77239-258 77239-260 77746-567
Alvelestat 98+%
Alvelestat
Precaución: for Research Use Only


Formula: C₂₅H₂₂F₃N₅O₄S
MDL Number: MFCD26960961
CAS Number: 848141-11-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty