Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoro-5-sulfamoylbenzoic acid 95%

Supplier: AMBEED, INC
Warning

A302794-1G A302794-250MG A302794-5G
77263-040EA 80 USD InStock
77263-040 77263-042 77263-044
2-Chloro-4-fluoro-5-sulfamoylbenzoic acid 95%
2-Chloro-4-fluoro-5-sulphamoylbenzoic acid
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₄S
MW: 253.6353 g/mol
MDL Number: MFCD01321357
CAS Number: 4793-24-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty