Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Piperidin-4-yloxy)isoquinolin-1(2H)-one 98+%

Supplier: AMBEED, INC
Warning

A303159-10MG A303159-1MG A303159-25MG A303159-5MG A303159-50MG
77250-810EA 35 USD InStock
77250-810 77250-812 77250-814 77250-816 77746-575
6-(Piperidin-4-yloxy)isoquinolin-1(2H)-one 98+%
6-(Piperidin-4-yloxy)isoquinolin-1(2H)-one
Precaución: for Research Use Only


Formula: C₁₄H₁₆N₂O₂
MDL Number: MFCD28335199
CAS Number: 923359-38-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty