Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A304597-1G A304597-250MG A304597-25G A304597-5G
77250-842EA 9 USD InStock
77250-842 77250-844 77250-846 77250-848
2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde 97%
2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₇H₆O₃S
MDL Number: MFCD01651766
CAS Number: 204905-77-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty