Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Benzyloxyindole 95% (contains 4% EtOH at maximum)

Supplier: AMBEED, INC
Warning

A305478-100G A305478-10G A305478-25G A305478-500G A305478-5G
77250-882EA 215 USD InStock
77250-882 77250-884 77250-886 77250-888 77250-890
5-Benzyloxyindole 95% (contains 4% EtOH at maximum)
5-Benzyloxyindole
Precaución: for Research Use Only


Formula: C₁₅H₁₃NO
MDL Number: MFCD00005676
CAS Number: 1215-59-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty