Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(7S,13R)-11-Fluoro-6,7,13,14-tetrahydro-7,13-dimethyl-1,15-etheno-1H-pyrazolo[4,3-f][1,4,8,10]benzoxatriazacyclotridecin-4(5H)-one ≥99%

Supplier: AMBEED, INC
Warning

A306917-10MG A306917-5MG A306917-250MG A306917-1G
77277-432EA 12 USD InStock
77277-432 77277-434 77616-334 77748-594
(7S,13R)-11-Fluoro-6,7,13,14-tetrahydro-7,13-dimethyl-1,15-etheno-1H-pyrazolo[4,3-f][1,4,8,10]benzoxatriazacyclotridecin-4(5H)-one ≥99%
(7S,13R)-11-Fluoro-6,7,13,14-tetrahydro-7,13-dimethyl-1,15-etheno-1H-pyrazolo[4,3-f][1,4,8,10]benzoxatriazacyclotridecin-4(5H)-one
Precaución: for Research Use Only


Formula: C₁₈H₁₈FN₅O₂
MDL Number: MFCD31544350
CAS Number: 1802220-02-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty