Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(methylthio)pyrazolo[1,5-a][1,3,5]triazine 97%

Supplier: AMBEED, INC
Warning

A308248-100MG A308248-250MG
77290-124EA 17 USD InStock
77290-124 77290-126
4-Chloro-2-(methylthio)pyrazolo[1,5-a][1,3,5]triazine 97%
4-Chloro-2-(methylthio)pyrazolo[1,5-a][1,3,5]triazine
Precaución: for Research Use Only


Formula: C₇H₆ClN₃S
MW: 199.66 g/mol
MDL Number: MFCD08275129
CAS Number: 54346-19-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty