Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Nitrotryptanthrin 98%

Supplier: AMBEED, INC
Warning

A309256-10MG A309256-1MG A309256-25MG A309256-5MG
77263-170EA 64 USD InStock
77263-170 77263-172 77263-174 77263-176
8-Nitrotryptanthrin 98%
8-Nitrotryptanthrin
Precaución: for Research Use Only


Formula: C₁₅H₇N₃O₄
MDL Number: MFCD00618023
CAS Number: 77603-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty