Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (S)-2-amino-4-carboxybutanoate hydrate 97%

Supplier: AMBEED, INC
Warning

A312257-100G A312257-25G A312257-500G
77239-440EA 31 USD InStock
77239-440 77239-442 77239-444
Potassium (S)-2-amino-4-carboxybutanoate hydrate 97%
Potassium (S)-2-amino-4-carboxybutanoate hydrate
Precaución: for Research Use Only


Formula: C₅H₈KNO₄·H₂O
MW: 203.23 g/mol
MDL Number: MFCD00216458
CAS Number: 6382-01-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty