Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-(4-Chlorophenyl)-2,2':6',2''-terpyridine 98%

Supplier: AMBEED, INC
Warning

A313556-100MG A313556-1G A313556-250MG
77239-450EA 13 USD InStock
77239-450 77239-452 77239-454
4'-(4-Chlorophenyl)-2,2':6',2''-terpyridine 98%
4'-(4-Chlorophenyl)-2,2':6',2''-terpyridine
Precaución: for Research Use Only


Formula: C₂₁H₁₄ClN₃
MDL Number: MFCD06796985
CAS Number: 89972-75-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty