Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

IWP-4 98%

Supplier: AMBEED, INC
Warning

A313997-10MG A313997-1MG A313997-25MG A313997-5MG
77263-250EA 86 USD InStock
77263-250 77263-252 77263-254 77263-256
IWP-4 98%
IWP-4
Precaución: for Research Use Only


Formula: C₂₃H₂₀N₄O₃S₃
MDL Number: MFCD04457754
CAS Number: 686772-17-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty