Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sitravatinib ≥98%

Supplier: AMBEED, INC
Warning

A314800-1MG A314800-25MG A314800-5MG
77239-462EA 22 USD InStock
77239-462 77239-464 77239-466
Sitravatinib ≥98%
Sitravatinib
Precaución: for Research Use Only


Formula: C₃₃H₂₉F₂N₅O₄S
MW: 629.68 g/mol
MDL Number: MFCD28502181
CAS Number: 1123837-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty