Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-(hydroxymethyl)benzonitrile 98%

Supplier: AMBEED, INC
Warning

A315064-100MG A315064-1G A315064-250MG A315064-5G
77263-258EA 22 USD InStock
77263-258 77263-260 77263-262 77263-264
2-Fluoro-4-(hydroxymethyl)benzonitrile 98%
2-Fluoro-4-(hydroxymethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₆FNO
MW: 151.1400 g/mol
MDL Number: MFCD08236861
CAS Number: 222978-02-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty