Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloro(2,6,10-dodecatriene-1,12-diyl)ruthenium(IV) 98%

Supplier: AMBEED, INC
Warning

A316334-100MG A316334-1G A316334-250MG
77251-066EA 70 USD InStock
77251-066 77251-068 77251-070
Dichloro(2,6,10-dodecatriene-1,12-diyl)ruthenium(IV) 98%
Dichloro(2,6,10-dodecatriene-1,12-diyl)ruthenium(IV)
Precaución: for Research Use Only


Formula: C₁₂H₁₈Cl₂Ru
MW: 334.2473 g/mol
MDL Number: MFCD08459354
CAS Number: 12170-97-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty