Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-([2,2':6',2''-Terpyridin]-4'-yl)phenol 97%

Supplier: AMBEED, INC
Warning

A316923-100MG A316923-1G A316923-250MG A316923-5G A316923-25G
77239-504EA 11 USD InStock
77239-504 77239-506 77239-508 77239-510 77746-609
4-([2,2':6',2''-Terpyridin]-4'-yl)phenol 97%
4-([2,2':6',2''-Terpyridin]-4'-yl)phenol
Precaución: for Research Use Only


Formula: C₂₁H₁₅N₃O
MDL Number: MFCD30478583
CAS Number: 89972-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty