Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Pyridin-4-yl)-1H-benzimidazole-6-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A318755-100MG A318755-250MG
77277-672EA 14 USD InStock
77277-672 77277-674
2-(Pyridin-4-yl)-1H-benzimidazole-6-carboxylic acid 97%
2-(Pyridin-4-yl)-1H-benzimidazole-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₉N₃O₂
MDL Number: MFCD06245477
CAS Number: 316833-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty