Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

JNJ-42041935 98+%

Supplier: AMBEED, INC
Warning

A319423-10MG A319423-1MG A319423-5MG
77239-554EA 58 USD InStock
77239-554 77239-556 77239-558
JNJ-42041935 98+%
JNJ-42041935
Precaución: for Research Use Only


Formula: C₁₂H₆ClF₃N₄O₃
MDL Number: MFCD25541711
CAS Number: 1193383-09-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty