Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-N-[(3-iodophenyl)methyl]-2',3'-O-(1-methylethylidene) Adenosine 95%

Supplier: AMBEED, INC
Warning

A322023-100MG A322023-1G A322023-250MG
77239-608EA 10 USD InStock
77239-608 77239-610 77239-612
2-Chloro-N-[(3-iodophenyl)methyl]-2',3'-O-(1-methylethylidene) Adenosine 95%
2-Chloro-N-[(3-iodophenyl)methyl]-2',3'-O-(1-methylethylidene) Adenosine
Precaución: for Research Use Only


Formula: C₂₀H₂₁ClIN₅O₄
MDL Number: MFCD30475782
CAS Number: 1000980-71-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty