Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,8,11-Trioxa-2-azatridecan-13-ol 97%

Supplier: AMBEED, INC
Warning

A322383-100MG A322383-1G A322383-250MG A322383-5G
77251-150EA 29 USD InStock
77251-150 77251-152 77251-154 77251-156
5,8,11-Trioxa-2-azatridecan-13-ol 97%
5,8,11-Trioxa-2-azatridecan-13-ol
Precaución: for Research Use Only


Formula: C₉H₂₁NO₄
MDL Number: MFCD29042347
CAS Number: 90430-59-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty