Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-3-thiopheneboronic acid 98%

Supplier: AMBEED, INC
Warning

A322720-100MG A322720-1G A322720-250MG A322720-5G
77290-310EA 10 USD InStock
77290-310 77290-312 77290-314 77290-316
4-Methyl-3-thiopheneboronic acid 98%
4-Methyl-3-thiopheneboronic acid
Precaución: for Research Use Only


Formula: C₅H₇BO₂S
MW: 141.99 g/mol
MDL Number: MFCD04039882
CAS Number: 177735-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty