Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Ethoxy-3-methoxyphenyl)methanamine 97%

Supplier: AMBEED, INC
Danger

A322789-100MG A322789-1G A322789-250MG
77277-720EA 19 USD InStock
77277-720 77277-722 77277-724
(4-Ethoxy-3-methoxyphenyl)methanamine 97%
(4-Ethoxy-3-methoxyphenyl)methanamine
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₂
MW: 181.23 g/mol
MDL Number: MFCD07754789
CAS Number: 93489-14-6
UN: 2735
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty