Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diamino-6-methoxy-1,3,5-triazine 98+%

Supplier: AMBEED, INC
Warning

A322822-1G A322822-250MG A322822-5G
77251-176EA 43 USD InStock
77251-176 77251-178 77251-180
2,4-Diamino-6-methoxy-1,3,5-triazine 98+%
2,4-Diamino-6-methoxy-1,3,5-triazine
Precaución: for Research Use Only


Formula: C₄H₇N₅O
MDL Number: MFCD00191340
CAS Number: 2827-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty