Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethylcyclopentanone 97%

Supplier: AMBEED, INC
Danger

A324989-100MG A324989-1G A324989-250MG A324989-5G
77251-194EA 5 USD InStock
77251-194 77251-196 77251-198 77251-200
2,2-Dimethylcyclopentanone 97%
2,2-Dimethylcyclopentanone
Precaución: for Research Use Only


Formula: C₇H₁₂O
MDL Number: MFCD00049164
CAS Number: 4541-32-6
UN: 1224
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty