Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dovitinib ≥99%

Supplier: AMBEED, INC
Warning

A325412-1MG A325412-50MG A325412-100MG
77290-352EA 11 USD InStock
77290-352 77290-354 77748-641
Dovitinib ≥99%
Dovitinib
Precaución: for Research Use Only


Formula: C₂₁H₂₁FN₆O
MDL Number: MFCD10565680
CAS Number: 405169-16-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty