Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4,6-dimethylnicotinonitrile 98%

Supplier: AMBEED, INC
Warning

A325511-10G A325511-1G A325511-250MG A325511-5G
77263-504EA 65 USD InStock
77263-504 77263-506 77263-508 77263-510
2-Methoxy-4,6-dimethylnicotinonitrile 98%
2-Methoxy-4,6-dimethylnicotinonitrile
Precaución: for Research Use Only


Formula: C₉H₁₀N₂O
MDL Number: MFCD00124716
CAS Number: 65515-39-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty