Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Spiro[fluorene-9,9'-xanthene] 97%

Supplier: AMBEED, INC
Warning

A329776-1G A329776-250MG A329776-5G
77239-744EA 15 USD InStock
77239-744 77239-746 77239-748
Spiro[fluorene-9,9'-xanthene] 97%
Spiro[fluorene-9,9'-xanthene]
Precaución: for Research Use Only


Formula: C₂₅H₁₆O
MDL Number: MFCD30342412
CAS Number: 159-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty