Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,5,5-Tetramethyldihydrofuran-3(2H)-one 97%

Supplier: AMBEED, INC
Danger

A329791-1G A329791-250MG A329791-25G
77251-284EA 11 USD InStock
77251-284 77251-286 77251-288
2,2,5,5-Tetramethyldihydrofuran-3(2H)-one 97%
2,2,5,5-Tetramethyldihydrofuran-3(2H)-one
Precaución: for Research Use Only


Formula: C₈H₁₄O₂
MDL Number: MFCD00009614
CAS Number: 5455-94-7
UN: 1224
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty