Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-1H-pyrazole-4-carboxamide 97%

Supplier: AMBEED, INC
Warning

A331926-10G A331926-1G A331926-250MG A331926-25G A331926-5G
77277-836EA 28 USD InStock
77277-836 77277-838 77277-840 77277-842 77277-844
3-Amino-1H-pyrazole-4-carboxamide 97%
3-Amino-1H-pyrazole-4-carboxamide
Precaución: for Research Use Only


Formula: C₄H₆N₄O
MDL Number: MFCD00082808
CAS Number: 5334-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty