Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl thieno[2,3-d]thiazole-5-carboxylate 97%

Supplier: AMBEED, INC
Warning

A333883-100MG A333883-1G A333883-250MG
77263-648EA 80 USD InStock
77263-648 77263-650 77263-652
Methyl thieno[2,3-d]thiazole-5-carboxylate 97%
Methyl thieno[2,3-d]thiazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₇H₅NO₂S₂
MDL Number: MFCD29127449
CAS Number: 1909316-58-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty